Learn More
Phenyl isocyanate, 98+%, Thermo Scientific™
Catalog Number p-7020747
Click to view available options
Quantity:
5 g
250 g
1000 g
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
103-71-9
C7H5NO
119.123
MFCD00001994
DGTNSSLYPYDJGL-UHFFFAOYSA-N
phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat
7672
CHEBI:53806
isocyanatobenzene
C1=CC=C(C=C1)N=C=O
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
103-71-9
C7H5NO
119.123
MFCD00001994
DGTNSSLYPYDJGL-UHFFFAOYSA-N
phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat
7672
CHEBI:53806
isocyanatobenzene
C1=CC=C(C=C1)N=C=O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Phenyl isocyanate is used with triethylamine in order to activate nitro groups to undergo 1,3-dipolar cycloaddition. It is also employed in the preparation of chemically modified cellulose paper. Further, it is used in the preparation of functionalized graphene oxide nanoplatelets. In addition to this, it reacts with ethyl alcohol to prepare ethyl alfa,gamma-diphenylallophanate and small amount of phenyl isocyanate dimer.
Solubility
Miscible with ether.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, water, moisture, alcohols, amines, strong bases and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point -33°C
Density 1.094
Boiling Point 62°C to 64°C (11 mmHg)
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.536
Quantity 5 g
UN Number UN2487
Beilstein 471391
Sensitivity Moisture sensitive
Merck Index 14,7296
Solubility Information Miscible with ether.
Formula Weight 119.12
Percent Purity ≥98%
Chemical Name or Material Phenyl isocyanate