Få mere at vide
Phenyl Di-p-tert-butylphenyl Phosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenyl Di-p-tert-butylphenyl Phosphate |
| CAS | 115-87-7 |
| Molekylær formel | C26 H31 O4 P |
| Mængde | 250 mg |
| Synonym | Phosphoric acid','bis[4-(1,1-dimethylethyl)phenyl] phenyl ester (9CI','ACI), Phenol','p-tert-butyl-','phenyl phosphate (2:1) (8CI), Phosphoric acid','bis(p-tert-butylphenyl) phenyl ester (6CI','7CI','8CI), Bis(4-tert-butylphenyl) phenyl phosphate, Bis(p-tert-butylphenyl) phenyl phosphate, NSC 44042, Phenyl di-p-tert-butylphenyl phosphate, Bis(4-tert-butylphenyl)phenyl phosphate |
| InChI formel | InChI=1S/C26H31O4P/c1-25(2,3)20-12-16-23(17-13-20)29-31(27,28-22-10-8-7-9-11-22)30-24-18-14-21(15-19-24)26(4,5)6/h7-19H,1-6H3 |
| SMIL | CC(C)(C)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccc(cc3)C(C)(C)C)cc1 |
| IUPAC navn | bis(4-tert-butylphenyl) phenyl phosphate |
| Molekylvægt (g/mol) | 438.5 |
| Formel vægt | 438.196 |