Learn More
Phenyl acrylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 937-41-7 |
| Molecular Formula | C9H8O2 |
| MDL Number | MFCD00048145 |
| Quantity | 500 g |
| SMILES | C=CC(OC1=CC=CC=C1)=O |
| Molecular Weight (g/mol) | 148.16 |
| Percent Purity | 97% |
| Chemical Name or Material | Phenyl acrylate |