Få mere at vide
Phenyl Acetate-d5, TRC
Artikelnummer. 30423627
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423627 100 mg
30417371 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423627 Leverandør Toronto Research Chemicals Leverandørnr. TRCP319152100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22705-26-6
Molekylær formel C8 2H5 H3 O2
Molekylvægt (g/mol) 141.18
Synonym Phenyl-d5 Acetic Acid Ester, Acetoxybenzene-d5, NSC 27795-d5, Phenol-d5 Acetate
IUPAC navn 2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid
SMIL [2H]c1c([2H])c([2H])c(CC(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale Phenyl Acetate-d5
Mængde 100 mg
InChI formel InChI=1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)/i1D,2D,3D,4D,5D
Formel vægt 141.0838