Få mere at vide
Phenyl 3,4-Di-O-acetyl-Alpha-O-benzyl-1-thio-Alpha-L-rhamnopyranoside, TRC
Artikelnummer. 30409825
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409825 50 mg
30414858 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30409825 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31970550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenyl 3,4-Di-O-acetyl-a-O-benzyl-1-thio-a-L-rhamnopyranosid
CAS 849938-20-1
Molekylær formel C23 H26 O6 S
Mængde 50 mg
Synonym Phenyl 3,4-Di-O-Acetyl-α-O-Benzyl-1-Thio-α-L-Rhamnopyranoside
InChI formel InChI=1S/C23H26O6S/c1-15-20(28-16(2)24)21(29-17(3)25)22(26-14-18-10-6-4-7-11-18)23(27-15)30-19-12-8-5-9-13-19/h4-13,15,20-23H,14H2,1-3H3/t15-,20-,21+,22+,23-/m0/s1
SMIL C[C@@H]1O[C@@H](Sc2ccccc2)[C@H](OCc3ccccc3)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC navn [(2S,3S,4R,5R,6S)-4-acetyloxy-2-methyl-5-phenylmethoxy-6-phenylsulfanyloxan-3-yl] acetate
Molekylvægt (g/mol) 430.51
Formel vægt 430.145