missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl(2-pyridyl)methanol, Mikromol™
Description
Phenyl(2-pyridyl)methanol, Mikromol
Specifications
Specifications
| Kemisk navn eller materiale | Phenyl(2-pyridyl)methanol |
| CAS | 14159-57-0 |
| Analyt- eller komponentnavne | Phenyl(2-pyridyl)methanol |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H11 N O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | NSC 241060, Phenyl(2-pyridinyl)methanol, Phenyl(2-pyridyl)methanol, Phenyl-2-pyridylcarbinol, alpha-Phenyl-2-pyridinemethanol, (+/-)-2-(alpha-Hydroxybenzyl)pyridine, (+/-)-2-Pyridylphenylmethanol, 2-(alpha-Hydroxybenzyl)pyridine |
| InChI formel | InChI=1S/C12H11NO/c14-12(10-6-2-1-3-7-10)11-8-4-5-9-13-11/h1-9,12,14H |
| Løsningstype | Neat |
| SMIL | OC(c1ccccc1)c2ccccn2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?