Få mere at vide
Phenyl-2,3,4,6-tetra-O-acetyl-Beta-D-galactopyranoside, TRC
Artikelnummer. 30418961
Skift visning
Mængde:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418961 25 g
30426120 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418961 Leverandør Toronto Research Chemicals Leverandørnr. TRCP33680025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenyl-2,3,4,6-tetra-O-acetyl-beta-D-galactopyranoside
CAS 2872-72-2
Molekylær formel C20H24O10
Mængde 25 g
Synonym Phenyl Tetra-O-acetyl-beta-D-galactopyranoside, NSC 173175
InChI formel InChI=1S/C20H24O10/c1-11(21)25-10-16-17(26-12(2)22)18(27-13(3)23)19(28-14(4)24)20(30-16)29-15-8-6-5-7-9-15/h5-9,16-20H,10H2,1-4H3/t16-,17+,18+,19-,20-/m1/s1
SMIL CC(=O)OC[C@H]1O[C@@H](Oc2ccccc2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC navn [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenoxyoxan-2-yl]methyl acetate
Molekylvægt (g/mol) 424.4
Formel vægt 424.1369