Få mere at vide
Phentolamine Mesilate EP Impurity A (as Oxalate), Mikromol™
Artikelnummer. 30049627
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30049627 25 mg
1 options
Artikelnummer. 30049627 Leverandør Mikromol Leverandørnr. MM1310.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phentolamine Mesilate EP Impurity A (as Oxalate)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2-Aminoethyl)-2-[(3-hydroxyphenyl)(4-methylphenyl)amino]acetamide Oxalate
CAS 2724727-53-9
Analyt- eller komponentnavne N-(2-Aminoethyl)-2-[(3-hydroxyphenyl)(4-methylphenyl)amino]acetamide Oxalate
Forsendelsestilstand Room Temperature
Molekylær formel C17 H21 N3 O2 . C2 H2 O4
Certificeringer/compliance ISO 9001
Synonym Phentolamine Mesilate Imp. A (EP), N-(2-Aminoethyl)-2-[(3-hydroxyphenyl)(4-methylphenyl)amino]acetamide Oxalate, Phentolamine Mesilate Impurity A as Oxalate, Phentolamine Impurity A as Oxalate
InChI formel InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24)
SMIL Cc1ccc(cc1)N(CC(=O)NCCN)c2cccc(O)c2.OC(=O)C(=O)O
IUPAC navn N-(2-aminoethyl)-2-(N-(3-hydroxyphenyl)-4-methylanilino)acetamide;oxalic acid
Molekylvægt (g/mol) 389.4
Mængde 25 mg
Formel vægt 389.1587 g/mol