Få mere at vide
Phentolamine-d4 Hydrochloride, TRC
Artikelnummer. 30410246
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30410246 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410246 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3025521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phentolamine-d4 Hydrochloride
CAS 1346599-65-2
Molekylær formel C17 D4 H15 N3 O . H Cl
Mængde 1 mg
Synonym 2-[N-(m-Hydroxyphenyl)-p-toluidinomethyl]imidazoline-d4 Hydrochloride, 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol-d4 Hydrochloride, DNP-d4, Regitine-d4 Hydrochloride
InChI formel InChI=1S/C17H19N3O.ClH/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H/i9D2,10D2;
SMIL Cl.[2H]C1([2H])NC(=NC1([2H])[2H])CN(c2ccc(C)cc2)c3cccc(O)c3
IUPAC navn 3-[4-methyl-N-[(4,4,5,5-tetradeuterio-1H-imidazol-2-yl)methyl]anilino]phenol;hydrochloride
Molekylvægt (g/mol) 321.84
Formel vægt 321.1546