missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phenthoat 1000 μg/ml i acetone, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenthoate |
| CAS | 2597-03-7 |
| Analyt- eller komponentnavne | Phenthoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H17 O4 P S2 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Acetic acid, mercaptophenyl-, ethyl ester O,O-dimethyl phosphorodithioate (6CI), Acetic acid, mercaptophenyl-, ethyl ester, S-ester with O,O-dimethyl phosphorodithioate (7CI), Acetic acid, mercaptophenyl-, ethyl ester, S-ester with O,O-dimethyl phosphorodithioate (8CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with ethyl mercaptophenylacetate (8CI), BAY 33051, Bayer 18,510, Bayer 18510, Cidemul, Cidial, Dhanusan 50, Dimephenthoate, ENT 23438, ENT 27386GC, Elsan, Fenthoate, L 561, Montecatini L 561, O,O-Dimethyl S-α-ethoxycarbonylbenzyl phosphorodithioate, Pap, Papthion, Phendal, Phenthoate, S-[α-(ethoxycarbonyl)benzyl] O,O-dimethyl phosphorodithioate, S-α-Ethoxycarbonylbenzyl dimethyl phosphorothiolothionate, Shaluosong, Tagson, Tsidial, Benzeneacetic acid, α-[(dimethoxyphosphinothioyl)thio]-, ethyl ester |
| InChI formel | InChI=1S/C12H17O4PS2/c1-4-16-12(13)11(10-8-6-5-7-9-10)19-17(18,14-2)15-3/h5-9,11H,4H2,1-3H3 |
| SMIL | CCOC(=O)C(SP(=S)(OC)OC)c1ccccc1 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?