Få mere at vide
Phenprocoumon-d5, TRC
Artikelnummer. 30420357
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30420357 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420357 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31882310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenprocoumon-d5
CAS 121513-38-0
Molekylær formel C18H11D5O3
Mængde 10 mg
Synonym 4-Hydroxy-3-[1-(phenyl-d5)propyl]-2H-1-Benzopyran-2-one, 3-(α-Ethylbenzyl)-4-hydroxy-coumarin-d5, (±)-Phenprocoumon-d5, 3-(1-Phenylpropyl)-4-hydroxycoumarin-d5, 3-(α-Ethylbenzyl)-4-hydroxycoumarin-d5, 3-(α-Phenylpropyl)-4-hydroxycoumarin-d5, 4-Hydroxy-2-oxo-3-(1-phenylpropyl)-2H-chromene-d5, BS 7565-d5, DL-3-(α-Ethylbenzyl)-4-hydroxycoumarin-d5, Falithrom-d5, Fencumar-d5, Liquamar-d5
InChI formel InChI=1S/C18H16O3/c1-2-13(12-8-4-3-5-9-12)16-17(19)14-10-6-7-11-15(14)21-18(16)20/h3-11,13,19H,2H2,1H3/i3D,4D,5D,8D,9D
SMIL O=C1C(C(C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H])CC)=C(O)C(C=CC=C3)=C3O1
Molekylvægt (g/mol) 285.35
Formel vægt 285.1413