missing translation for 'onlineSavingsMsg'
Få mere at vide

Phenprobamate, TRC

Artikelnummer. 30410642
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30410642 10 g
30417565 50 g
30422406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410642 Supplier Toronto Research Chemicals Supplier No. TRCP31885810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-31-4
Molekylær formel C10 H13 N O2
Molekylvægt (g/mol) 179.22
Synonym Benzenepropanol, 1-carbamate, 1-Propanol, 3-phenyl-, carbamate (8CI), Benzenepropanol, carbamate (9CI), Carbamic acid, 3-phenylpropyl ester (6CI,7CI), 3-Phenyl-1-propanol carbamate, 3-Phenylpropyl carbamate, Ansepron, Eirenal, Extacol, Gamaquil, MH 532, NSC 44682, NSC 50538, NSC 64270, Palmita, Phenprobamate, Proformiphen, Quamaquil, Spantol, Tranquil, γ-Phenylpropyl carbamate
IUPAC navn 3-phenylpropyl carbamate
SMIL NC(=O)OCCCc1ccccc1

Specifications

Kemisk navn eller materiale Phenprobamate
Mængde 10 g
InChI formel InChI=1S/C10H13NO2/c11-10(12)13-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H2,11,12)
Formel vægt 179.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.