Få mere at vide
Phenoxypropazine Dihydrochloride, TRC
Artikelnummer. 30418372
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418372 1 g
30414857 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418372 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3186651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1803608-13-0
Molekylær formel C9 H14 N2 O . 2 Cl H
Molekylvægt (g/mol) 239.14
Synonym Phenoxypropazine dihydrochloride, Phenoxypropazine salt, Phenoxypropazine
IUPAC navn 1-phenoxypropan-2-ylhydrazine;dihydrochloride
SMIL Cl.Cl.CC(COc1ccccc1)NN
Tekniske data
Kemisk navn eller materiale Phenoxypropazine Dihydrochloride
Mængde 1 g
InChI formel InChI=1S/C9H14N2O.2ClH/c1-8(11-10)7-12-9-5-3-2-4-6-9;;/h2-6,8,11H,7,10H2,1H3;2*1H
Formel vægt 238.064