Få mere at vide
Phenoxymethylpenilloic Acid (Mixture of Diastereomers), TRC
Artikelnummer. 30006568
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006568 10 mg
30011023 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006568 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29825010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4847-29-4
Molekylær formel C15 H20 N2 O4 S
Molekylvægt (g/mol) 324.4
Synonym 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-, 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[(2-phenoxyacetamido)methyl]- (6CI,7CI,8CI), 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[[(phenoxyacetyl)amino]methyl]- (9CI), 5,5-Dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-4-thiazolidinecarboxylic acid, Penilloic acid V, Penilloic acid, phenoxymethyl-, Phenoxymethylpenilloic acid, 5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic acid
IUPAC navn 5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-1,3-thiazolidine-4-carboxylic acid
SMIL CC1(C)SC(CNC(=O)COc2ccccc2)NC1C(=O)O
Tekniske data
Kemisk navn eller materiale Phenoxymethylpenilloic Acid
Mængde 10 mg
InChI formel InChI=1S/C15H20N2O4S/c1-15(2)13(14(19)20)17-12(22-15)8-16-11(18)9-21-10-6-4-3-5-7-10/h3-7,12-13,17H,8-9H2,1-2H3,(H,16,18)(H,19,20)
Formel vægt 324.1144