Få mere at vide
Phenolphthalein Disodium Salt (>90%), TRC
Artikelnummer. 30428584
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428584 250 mg
30418225 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428584 Leverandør Toronto Research Chemicals Leverandørnr. TRCP318033250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenolphthalein Disodium Salt
CAS 518-51-4
Molekylær formel C20H12Na2O4
Mængde 250 mg
InChI formel InChI=1S/C20H14O4.2Na/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20;;/h1-12,21-22H;;/q;2*+1/p-2
SMIL [Na+].[Na+].[O-]c1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc([O-])cc4
IUPAC navn disodium;4-[1-(4-oxidophenyl)-3-oxo-2-benzofuran-1-yl]phenolate
Molekylvægt (g/mol) 362.29
Formel vægt 362.0531