Få mere at vide
Phenol-d6, CDN
Artikelnummer. 30310574
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30310574 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310574 Leverandør CDN Leverandørnr. CDND00295G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenol D6
CAS 13127-88-3
Molekylær formel C6 2H6 O
Mængde 5 g
Synonym Phenol-D6, Phen-2,3,4,5,6-d5-ol-d, Phenol-d6 (7 CI,8 CI,9 CI), Hexadeuteriophenol
InChI formel InChI=1 S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7 H/i1D,2 D,3 D,4 D,5 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-deuteriooxybenzene
Molekylvægt (g/mol) 100.15
Formel vægt 100.0795 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity