Få mere at vide
Phenol D6 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30313816
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30313816 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313816 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA16025200AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenol D6
CAS 13127-88-3
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C6 2H6 O
Mængde 1 mL
Synonym Phenol-D6, Phen-2,3,4,5,6-d5-ol-d, Phenol-d6 (7CI,8CI,9CI), Hexadeuteriophenol
InChI formel InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,2D,3D,4D,5D/hD
SMIL [2H]Oc1c([2H])c([2H])c([2H])c([2H])c1[2H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-deuteriooxybenzene
Molekylvægt (g/mol) 100.15
Formel vægt 100.0795 g/mol