Få mere at vide
Pheniramine Maleate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pheniramine Maleate |
| CAS | 132-20-7 |
| Analyt- eller komponentnavne | Pheniramine Maleate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H20 N2 . C4 H4 O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Brompheniramine Maleate Imp. C (EP), Brompheniramine Imp. C (EP), Dexchlorpheniramine Maleate Imp. A (EP), Dexchlorpheniramine Imp. A (EP), (3RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine Maleate, Pheniramine Maleate, Dexchlorpheniramine Maleate Imp. A (EP) as Maleate, Brompheniramine Maleate Imp. C (EP) as Maleate, N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (Z)-2-butenedioate (1:1), Prophenpyridamine maleate, 2-[alpha-[2-(Dimethylamino)ethyl]benzyl]pyridine maleate (1:1), 1-Phenyl-1-(2-pyridyl)-3-dimethaminopropane maleinate, 1-Phenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, 2-[alpha-[2-(Dimethylamino)ethyl]benzyl]pyridine bimaleate, Daneral, Inhiston, Metron, Naphcon A, PM 241, Pheniramine hydrogen maleate, Pheniramine maleate, Trimeton, Trimetose, Brompheniramine Maleate Impurity C as Maleate, Dexchlorpheniramine Maleate Impurity A as Maleate, Dexchlorpheniramine Impurity A as Maleate |
| InChI formel | InChI=1S/C16H20N2.C4H4O4/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16;5-3(6)1-2-4(7)8/h3-10,12,15H,11,13H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Løsningstype | Neat |
| SMIL | CN(C)CCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| Vis mere |