Få mere at vide
Phenglutarimide Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenglutarimide Hydrochloride |
| CAS | 1674-96-0 |
| Analyt- eller komponentnavne | Phenglutarimide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H24 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2,6-Piperidinedione, 3-[2-(diethylamino)ethyl]-3-phenyl-, hydrochloride (1:1), 2,6-Piperidinedione, 3-[2-(diethylamino)ethyl]-3-phenyl-, monohydrochloride (9CI), Glutarimide, 2-[2-(diethylamino)ethyl]-2-phenyl-, hydrochloride (7CI), Glutarimide, 2-[2-(diethylamino)ethyl]-2-phenyl-, monohydrochloride (8CI), Aturbal, Aturban, Aturban hydrochloride, Aturbane, Phenglutarimide hydrochloride |
| InChI formel | InChI=1S/C17H24N2O2.ClH/c1-3-19(4-2)13-12-17(14-8-6-5-7-9-14)11-10-15(20)18-16(17)21;/h5-9H,3-4,10-13H2,1-2H3,(H,18,20,21);1H |
| Løsningstype | Neat |
| SMIL | Cl.CCN(CC)CCC1(CCC(=O)NC1=O)c2ccccc2 |
| Vis mere |