Få mere at vide
Phenformin Hydrochloride, TRC
Artikelnummer. 30423794
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423794 5 g
30423427 10 g
30410304 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423794 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2969005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 834-28-6
Molekylær formel C10 H15 N5 . Cl H
Molekylvægt (g/mol) 241.72
Synonym Imidodicarbonimidic diamide, N-(2-phenylethyl)-, hydrochloride (1:1), Biguanide, 1-phenethyl-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N-(2-phenylethyl)-, monohydrochloride (9CI), 1-(2-Phenylethyl)biguanide hydrochloride, 1-Phenethylbiguanide hydrochloride, 1-β-Phenylethylbiguanide hydrochloride, Azucaps, DBI-TD, Debeone DT, Debinyl, Dibein, Dipar, Feguanide, Glucopostin, Insoral, Lentobetic, Meltrol, N-Phenethylbiguanide hydrochloride, Normoglucina, N'-β-Phenylethylbiguanide-hydrochloride, Phenethylbiguanide hydrochloride, Phenformin hydrochloride, α-Phenethylbiguanide hydrochloride
IUPAC navn 1-carbamimidoyl-3-(2-phenylethyl)guanidine;hydrochloride
SMIL Cl.NC(=N)NC(=N)NCCc1ccccc1
Tekniske data
Kemisk navn eller materiale Phenformin Hydrochloride
Renhedsgrad noter HPLC
Mængde 5 g
InChI formel InChI=1S/C10H15N5.ClH/c11-9(12)15-10(13)14-7-6-8-4-2-1-3-5-8;/h1-5H,6-7H2,(H6,11,12,13,14,15);1H
Formel vægt 241.1094
Procent renhed >95%