Få mere at vide
Phenformin hydrochloride, MedChemExpress
Artikelnummer. 16489587
Encompass_Preferred
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16489587 5 g 5g
16471609 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16489587 Leverandør Medchem Express Leverandørnr. HY16397A
MedChemExpress Phenformin hydrochloride is an anti-diabetic drug from the biguanide class, can activate AMPK activity.
Phenformin hydrochloride is an anti-diabetic drug from the biguanide class, can activate AMPK activity.
Tekniske data
Kemisk navn eller materiale Phenformin hydrochloride
Fysisk form Powder
CAS 834-28-6
Farve White
Mængde 5 g
Renhedsgrad noter Research
Molekylær formel C10H16ClN5
Til brug med (applikation) Cancer-Kinase/protease
Synonym Phenethylbiguanide hydrochloride
Opløselighedsinformation H2O : 12.5 mg/mL (51.71 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.14 mM; ultrasonic and adjust pH to 2 with 1M HCl) ∣Ethanol : < 1 mg/mL (insoluble) ∣DMF : < 1 mg/mL (insoluble)
SMIL NC(NC(NCCC1=CC=CC=C1)=N)=N.Cl
Molekylvægt (g/mol) 241.72
Formel vægt 241.72
Procent renhed 98.0%