Få mere at vide
Phenethyl Alcohol-d5, TRC
Artikelnummer. 30428322
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30428322 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428322 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2960021G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenethyl Alcohol-d5
CAS 35845-63-7
Molekylær formel C8 D5 H5 O
Mængde 1 g
Synonym Benzeneethanol-d5, (2-Hydroxyethyl)benzene-d5, 2-Phenethanol-d5, 2-Phenyl-1-ethanol-d5, 2-Phenylethanol-d5, Phenylethyl Alcohol-d5, β-Phenethanol-d5, β-Phenethyl Alcohol-d5, β-Phenethylol-d5, β-Phenylethanol-d5, β-Phenylethyl Alcohol-d5, NSC 406252-d5, 2-Phenyl-d5-ethanol
InChI formel InChI=1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(CCO)c([2H])c1[2H]
IUPAC navn 2-(2,3,4,5,6-pentadeuteriophenyl)ethanol
Molekylvægt (g/mol) 127.2
Formel vægt 127.1045