Få mere at vide
Phenelzine Sulfate, Mikromol™
Beskrivelse
Phenelzine Sulfate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenelzine Sulfate |
| CAS | 156-51-4 |
| Analyt- eller komponentnavne | Phenelzine Sulfate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H12 N2 . H2 O4 S |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Hydrazine, (2-phenylethyl)-, sulfate (1:1), Hydrazine, phenethyl-, sulfate (1:1) (8CI), (2-Phenylethyl)hydrazine dihydrogen sulfate, 2-Phenethylhydrazine sulfate, Estinerval, Fenelzin, Kalgan, Mao-rem, NSC 170957, Nardelzine, Nardil, Phenalzine dihydrogen sulfate, Phenelzine acid sulfate, Phenelzine bisulfate, Phenelzine dihydrogen sulfate, Phenelzine hydrogen sulfate, Phenelzine sulfate, Phenethylhydrazine sulfate, Stinerval, W 1544A, (beta-Phenylethyl)hydrazine dihydrogen sulfate, (beta-Phenylethyl)hydrazine sulfate |
| InChI formel | InChI=1S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10H,6-7,9H2;(H2,1,2,3,4) |
| SMIL | NNCCc1ccccc1.OS(=O)(=O)O |
| IUPAC navn | 2-phenylethylhydrazine;sulfuric acid |
| Vis mere |