Få mere at vide
Phenelzine-d5 Sulfate (phenyl-d5), CDN
Artikelnummer. 30318599
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318599 0.05 g
30314886 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318599 Leverandør CDN Leverandørnr. CDND58930.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenelzine-D5 (phenyl-D5) Sulfate
CAS 1219798-40-9
Molekylær formel C8 D5 H7 N2 . H2 O4 S
Mængde 0.05 g
Synonym [2-(Phenyl-d5)ethyl]hydrazine sulfate, 1-(2-Phenethyl)hydrazine-d5 sulfate, 1-Hydrazino-2-(phenyl-d5)ethane sulfate, Phenelzine-d5 sulfate, Phenelzine-D5 (phenyl-D5) sulfate
InChI formel InChI=1 S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10 H,6-7,9H2;(H2,1,2,3,4)/i1D,2 D,3 D,4 D,5 D;
SMIL [2 H]c1c([2 H])c([2 H])c(CCNN)c([2 H])c1[2 H].OS(=O)(=O)O
IUPAC navn 2-(2,3,4,5,6-pentadeuteriophenyl)ethylhydrazine;sulfuric acid
Molekylvægt (g/mol) 239.304
Formel vægt 239.099 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity