Få mere at vide
Phenbenzamine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phenbenzamine Hydrochloride |
| CAS | 2045-52-5 |
| Analyt- eller komponentnavne | Phenbenzamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H22 N2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,2-Ethanediamine, N1,N1-dimethyl-N2-phenyl-N2-(phenylmethyl)-, hydrochloride (1:1), 1,2-Ethanediamine, N,N-dimethyl-N'-phenyl-N'-(phenylmethyl)-, monohydrochloride (9CI), Ethylenediamine, N-benzyl-N',N'-dimethyl-N-phenyl-, hydrochloride (7CI,8CI), Antergan hydrochloride, N-Benzyl-N',N'-dimethyl-N-phenylethylenediamine hydrochloride, Phenbenzamine chloride, Phenbenzamine hydrochloride |
| InChI formel | InChI=1S/C17H22N2.ClH/c1-18(2)13-14-19(17-11-7-4-8-12-17)15-16-9-5-3-6-10-16;/h3-12H,13-15H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CN(C)CCN(Cc1ccccc1)c2ccccc2 |
| Vis mere |