missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phenazopyridine hydrochloride, MedChemExpress
Description
Phenazopyridine hydrochloride is a chemical, which has a local analgesic effect, often used to alleviate the pain, irritation, discomfort, or urgency caused by urinary tract infections, surgery, or injury to the urinary tract.
Specifications
Specifications
| Kemisk navn eller materiale | Phenazopyridine hydrochloride |
| CAS | 136-40-3 |
| Farve | Red |
| Renhedsgrad noter | Research |
| Molekylær formel | C11H12ClN5 |
| Mængde | 100 mg |
| Opløselighedsinformation | DMSO : 25 mg/mL (100.12 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.00 mM; ultrasonic and warming and heat to 60°C) |
| SMIL | NC1=NC(N)=CC=C1/N=N/C2=CC=CC=C2.[H]Cl |
| Molekylvægt (g/mol) | 249.7 |
| Formel vægt | 249.7 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?