missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrenequinone, 95%, Thermo Scientific Chemicals

Catalog Number p-180813
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
84-11-7
C14H8O2
208.22
MFCD00001163
YYVYAPXYZVYDHN-UHFFFAOYSA-N
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
6763
CHEBI:37454
phenanthrene-9,10-dione
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O
This item is not returnable. View return policy
84-11-7
C14H8O2
208.22
MFCD00001163
YYVYAPXYZVYDHN-UHFFFAOYSA-N
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
6763
CHEBI:37454
phenanthrene-9,10-dione
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 201°C to 211°C
Color Brown-Orange
Boiling Point 360°C
Flash Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07, 796
Fieser 01,827; 02,317
Merck Index 15, 7327
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in glacial acetic acid,soluble in benzene,ether and hot alchol
Formula Weight 208.22
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Phenanthrenequinone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Phenanthrenequinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'