Learn More
Phenanthrene-d10, TRC
Product Code. 30421328
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30421328 250 mg 250mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421328 Supplier Toronto Research Chemicals Supplier No. TRCP294801250MG
 Restricted Item
Forventet forsendelse: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenanthrene D10
CAS 1517-22-2
Purity Grade Notes HPLC
Molecular Formula C14 2H10
Quantity 250 mg
Synonym Phenanthrene-1,2,3,4,5,6,7,8,9,10-d10, Phenanthrene-d10 (6CI,7CI,8CI,9CI), 1,2,3,4,5,6,7,8,9,10-Decadeuteriophenanthrene, Perdeuterated phenanthrene, Perdeuteriophenanthrene, Perdeuterophenanthrene, [2H10]Phenanthracene, Phenanthrene D10
InChI Formula InChI=1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMILES [2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c([2H])c([2H])c([2H])c([2H])c23
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuteriophenanthrene
Molecular Weight (g/mol) 188.29
Formula Weight 188.141
Percent Purity >95