missing translation for 'onlineSavingsMsg'
Learn More

Phenamacril, TRC

Product Code. 30416286
Change view
Click to view available options
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30416286 50 mg
30415636 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416286 Supplier Toronto Research Chemicals Supplier No. TRCP29478550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39491-78-6
Molekylær formel C12 H12 N2 O2
Molekylvægt (g/mol) 216.24
Synonym 2-Propenoic acid, 3-amino-2-cyano-3-phenyl-, ethyl ester, (2Z)-, 2-Propenoic acid, 3-amino-2-cyano-3-phenyl-, ethyl ester, (Z)- (9CI), Ethyl (2Z)-3-amino-2-cyano-3-phenylprop-2-enoate, Ethyl (Z)-3-amino-2-cyano-3-phenylprop-2-enoate, Phenamacril, Phenamacryl
IUPAC navn ethyl (Z)-3-amino-2-cyano-3-phenylprop-2-enoate
SMIL CCOC(=O)\C(=C(/N)\c1ccccc1)\C#N

Specifications

Kemisk navn eller materiale Phenamacril
Mængde 50 mg
InChI formel InChI=1S/C12H12N2O2/c1-2-16-12(15)10(8-13)11(14)9-6-4-3-5-7-9/h3-7H,2,14H2,1H3/b11-10-
Formel vægt 216.0899
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.