Få mere at vide
Phenacetin-d5, TRC
Artikelnummer. 30410399
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30410399 25 mg -
30426918 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410399 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29458225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenacetin-d5
CAS 69323-74-6
Molekylær formel C10 2H5 H8 N O2
Mængde 25 mg
Synonym Acetamide','N-[4-(ethoxy-d5)phenyl]- (9CI), N-[4-(Ethoxy-d5)phenyl]acetamide, Phenacetin-d5, Phenacetin-D5 (ethoxy-D5), p-Acetophenetidide-d5, 4-(Acetylamino)phenetole-d5, 4-Ethoxyacetanilide-d5, N-Acetyl-4-(ethoxy-d5)aniline, N-Acetyl-p-phenetidine-d5
InChI formel InChI=1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1ccc(NC(=O)C)cc1
IUPAC navn N-[4-(1,1,2,2,2-pentadeuterioethoxy)phenyl]acetamide
Molekylvægt (g/mol) 184.25
Formel vægt 184.126