Få mere at vide
Phenacetin-d3, TRC
Artikelnummer. 30411914
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30411914 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411914 Leverandør Toronto Research Chemicals Leverandørnr. TRCP294583100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenacetin-d3
CAS 60902-27-4
Molekylær formel C10H10D3NO2
Mængde 100 mg
Synonym N-(4-Ethoxyphenyl)acetamide-d3, p-Acetophenetidide-d3, 4-(Acetyl-d3-amino)phenetole, 4-Ethoxy-1-acetyl-d3-aminobenzene, 4-Ethoxyacet-d3-anilide, 4'-Ethoxyacet-d3-anilide, Aceto-d3-4-phenetidine, Aceto-d3-phenetidin, Aceto-d3-phenetidine, Aceto-d3-phenetin, Acet-d3-phenetidin, Fenidina-d3, Fenina-d3, Kalmin-d3, N-Acetyl-d3-4-ethoxyaniline, N-Acetyl-d3-p-ethoxyaniline, N-Acetyl-d3-p-phenetidine, NSC 7651-d3, Pertonal-d3, Phenacetine-d3, Phenazetin-d3, Phenedina-d3, Phenidin-d3, Phenin-d3, p-Ethoxyacet-d3-anilide
InChI formel InChI=1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)/i2D3
SMIL O=C(C([2H])([2H])[2H])NC1=CC=C(OCC)C=C1
Molekylvægt (g/mol) 182.23
Formel vægt 182.1135