Få mere at vide
Phenacemide, LoGiCal
Artikelnummer. 30463106
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30463106 5 mg
1 options
Artikelnummer. 30463106 Leverandør LoGiCal Leverandørnr. LGCFOR1431.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenacemide
CAS 63-98-9
Analyt- eller komponentnavne Phenacemide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H10 N2 O2
Certificeringer/compliance ISO 9001
Synonym N-(Aminocarbonyl)benzeneacetamide, (Phenylacetyl)urea, Cetylureum, Epiclase, Fenacemid, NSC 39458, Neophedan, Phacetur, Phenacalum, Phenacemide, Phenacetur, Phenacetylcarbamide, Phenacetylurea, Phenicarb, Phenutal, Phetylureum, alpha-Phenylacetylurea
InChI formel InChI=1S/C9H10N2O2/c10-9(13)11-8(12)6-7-4-2-1-3-5-7/h1-5H,6H2,(H3,10,11,12,13)
Løsningstype Neat
SMIL NC(=O)NC(=O)Cc1ccccc1
IUPAC navn N-carbamoyl-2-phenylacetamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 178.19
Mængde 5 mg
Formel vægt 178.0742 g/mol