missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ PharMed™ BPT L/S™ 2-Stop Pump Tubing

Protect your light-sensitive samples

Brand:  Masterflex™ 96114-35

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11856683

  • 585.00 DKK / Pack of 4


Estimated Shipment
29-07-2021
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable
  • For use with Masterflex™ multichannel pump heads.
Specifications

Specifications

Beige
35 psi
Yes
PharMed BPT
0.38 m
Cell and Tissue Culture Work, Pharmaceutical, Light-Sensitive Samples
4
2600 mL/min
100°C
660 mm Hg
7.9 mm
1.25 ft.
Stopped Pump Tubing
-20°C
35
3/8 in.
Videos
SDS
Documents