Få mere at vide
Christeyns™ Phago'Derm Sensitive SPS sæbe
Volumen (metrisk):
500 ml
1000 ml
Mængde:
12 flasker
12 luftfrit monterede pumpeflasker
20 firkantede pumpeflasker
Beskrivelse
- Does not dry out the skin, even with frequent use
- Hypoallergenic (formulated to minimize allergic reactions)
- Suitable for washing children under 36 months and pregnant/breastfeeding women
- Suitable for patient washing and preoperative showering
- Good cleaning power
- High rinsability
- Hydrating potential for 30 min.
Tekniske data
Tekniske data
| Produkttype | Hand Soap |
| Farve | Colorless |
| Form | Foam |
| Mængde | 20 firkantede pumpeflasker |
| Volumen (metrisk) | 500 ml |