missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Blue Litmus Paper Reel
Provides a general test for acidic or alkaline reactions. Fisherbrand™ Blue Litmus Paper Reels are comprised of completely impregnated indicator paper.
Brand: Fisherbrand
Additional Details : Weight : 0.06016kg
Description
- Color change from blue to red
- Store in a cool, dry place
- Avoid exposure to sunlight and moisture
- Serves in neutralization reactions to check whether the change from acid to alkaline or vice versa has taken place
Specifications
Blue to Red | |
Reel | |
1 Each | |
5000 mm |
8 to 5 | |
Determination of acidic and alkaline reaction | |
pH Paper | |
7 mm |