Få mere at vide
SI Analytics™ pH-kombinations mikro-elektrode
Shop alle SI Analytics produkter
Konnektortyper:
BNC/RCA
DIN
DIN og 4 mm bananstik
DIN, 4 mm bananstik, 4 mm Banana Plug
Fast kabel
Stikhoved
Type:
pH-kombinationselektrode
pH-mikrokombinationselektrode
membran:
Cylinder
Flasker
Kugle
Sfære
Spyd
Diameter (metrisk):
12/3 mm
3 mm
5 mm
Pakningsstørrelse:
Hver
Beskrivelse
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Tekniske data
Tekniske data
| Konnektortyper | DIN, 4 mm bananstik, 4 mm Banana Plug |
| Knudepunkt | Platinum Diaphragm |
| Materiale | A-Glass |
| pH-område | 0 to 14 |
| Type | pH-kombinationselektrode |
| Intern reference | Silamid™ |
| membran | Sfære |
| Model- | A 157 1M-Din-Id |
| Temperaturområde (metrisk). | -5°C to +100°C |
| Længde (metrisk) | 40/130 mm |
| Vis mere |