Learn More
pH Buffer Solution pH 7, Fisher Chemical™
Shop All Reagecon Products
Click to view available options
Quantity:
10 L
Unit Size:
10 L
Specifications
Specifications
Color | Undesignated |
pH | 7 |
Quantity | 10 L |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Product Title
pH Buffer Solution pH 7, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction