Få mere at vide
pH Buffer pH 5, Fisher Chemical™
Shop alle Reagecon produkter
Mængde:
5 L
Pakningsstørrelse:
5 L
Beskrivelse
Mercury free buffers at 25°C (pH4 and pH10 are coloured) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Tekniske data
Tekniske data
| pH | 5 |
| Mængde | 5 L |