Learn More
Invitrogen™ PGRPs/PGLYRP1 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHPGLYRP1X5
Additional Details : Weight : 0.33820kg
Description
The Human PGRPs/PGLYRP1 ELISA quantitates Hu PGRPs in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PGRPs. Principle of the method The Human PGRPs solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The primary immune recognition is based on structures common among invading pathogens. Bacterial surface molecules, such as lipopolysaccharide (LPS) and peptidoglycan (PGN), are known to elicit immune reactions ranging from cytokine release to fever. Recently, a family of proteins called peptidoglycan recognition protein (PGRP) has been identified in mouse and human that binds to peptidoglycans expressed on Gram-positive bacteria. Peptidoglycan (PGN) is an essential cell wall component of virtually all bacteria (1,2) and, thus, it is an excellent target for recognition by the eukaryotic innate immune system. The PGRPs (PGRP-L, PGRP-S, PGRP-Ia, and PGRP-Ib) define a new family of human pattern recognition molecules (3). PGRP-L is primarily expressed in the liver. Although liver is not considered a primary immune organ, liver participates in host defenses by producing acute phase proteins (by hepatocytes) in response to infections and by clearing microorganisms from blood (4). PGRP-S is present in neutrophils and inhibits growth of Gram-positive bacteria and, therefore, may function as a neutrophil antibacterial protein (5). However, PGRP-S may have another, as yet unidentified function because in humans it is expressed in the bone marrow 50-100 times higher than in neutrophils.Specifications
O75594 | |
6 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
8993 | |
<12% | |
Stop Solution, SAV-HRP, Pre-coated 96 well plates, Biotinylated Detection Antibody, Adhesive Plate Covers, Chromogen, Wash Buffer, Standard, Assay Diluent Concentrate | |
5 x 96 Tests | |
Inflammation | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
6.14 pg/mL to 1500 pg/mL | |
6 pg/mL to 1500 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA pair antibody detects human PGRPs, less than 0.05% cross-reactivity with human PGRP-1 beta and mouse PGRPs is observed. | |
Human | |
Colorimetric Microplate Reader | |
PGLYRP1 | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L | |
SBBI68, PGLYRP, PGRP, PGRP-S, PGRPS, TAG7, TNFSF3L | |
1 hr. 20 min. |