missing translation for 'onlineSavingsMsg'
Få mere at vide

PGC1 alpha Antibody, Allophycocyanin, Novus Biologicals™

Artikelnummer. 18282235 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
18282235 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18282235 Leverandør Novus Biologicals Leverandørnr. NBP104676APC

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

PGC1 alpha Polyclonal antibody specifically detects PGC1 alpha in Human, Mouse, Rat, Porcine, Goat, Hamster, Sheep, Squirrel samples. It is validated for Flow Cytometry, Flow (Intracellular), KnockDown
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PGC1 alpha
Applikationer Flow Cytometry, KnockDown
Klassifikation Polyclonal
Konjugeret APC
Fortynding Flow Cytometry, Flow (Intracellular), Knockout Validated
Formulering PBS
Gene Alias LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha
Værtsarter Rabbit
Immunogen This PGC1 alpha Antibody was developed against a recombinant protein made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot# Q9UBK2].
Oprensningsmetode Immunogen affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Alzheimers Research, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Epigenetics, Lipid and Metabolism, Mitochondrial Mediated Pathway, mTOR Pathway, Neuroscience, Transcription Factors and Regulators
Primær eller sekundær Primary
Gen-id (Entrez) 10891
Målarter Human, Mouse, Rat, Pig, Goat, Hamster, Sheep, Squirrel
Indhold og opbevaring Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.