missing translation for 'onlineSavingsMsg'
Få mere at vide

PFN1 Mouse anti-Human, CoraLite 594, Clone: 3A12E8, Proteintech

Artikelnummer. 17264826 Shop alle Proteintech produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17264826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17264826 Leverandør Proteintech Leverandørnr. CL59467390100UL
 Begrænset vare
På lager
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Profilin-1 (PFN1) plays an important role in the control of actin dynamics, and could represent an important therapeutic target in several diseases. PFN1 is identified as a huntingtin aggregation inhibitor, and may serves as a tumor-suppressor. PFN1 is crucial for the conversion of monomeric (G)-actin to filamentous (F)-actin. Amyotrophic lateral sclerosis (ALS) is a late-onset neurodegenerative disorder resulting from motor neuron death. Cells expressing PFN1 mutants contain ubiquitinated, insoluble aggregates that in many cases contain the ALS-associated protein TDP-43. Recently, PFN1 is a potential biomarker for bladder cancer aggressiveness and may be of great clinical importance.

Tekniske data

Antigen PFN1
Applikationer Immunofluorescence
Klassifikation Monoclonal
Klon 3A12E8
Koncentration 991 μg/mL
Konjugeret CoraLite 594
Værtsarter Mouse
Mængde 100 μL
Primær eller sekundær Primary
Gen-id (Entrez) 5216
Målarter Human, Mouse, Rat
Form Liquid
Isotype IgG2b
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.