Få mere at vide
Tocris Bioscience™ PFI 4
Potent and selective BRPF1 bromodomain inhibitor
Brand:  Tocris Bioscience™ 5576/10
Encompass
Produktkode 15756936
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
PFI 4 is a potent and selective BRPF1B bromodomain inhibitor (IC50 = 172 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of other bromodomains including BRPF2 (BRD1), BRPF3 and BRD4. Disrupts BRPF1 binding to histone H3.3. Cell permeable. Inhibits osteoclastogenesis.
Tekniske data
Tekniske data
PFI 4
N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide
Soluble to 50mM in DMSO and to 10mM in ethanol
Bioactive Small Molecule
900305-37-5
10 mg
Solid
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
Research Use Only. Not For Human or Veterinary Use