Få mere at vide
Tocris Bioscience™ PF 915275
GREENER_CHOICE
Artikelnummer. 18075522
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18075522 50 mg
18045452 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18075522 Leverandør Tocris Bioscience™ Leverandørnr. 3291/50
Potent and selective 11β-hydroxysteroid dehydrogenase inhibitor
Potent and selective 11β-hydroxysteroid dehydrogenase type 1 (11βHSD1) inhibitor (Ki = 2.3 nM) that displays little activity at 11βHSD2 (1.5% inhibition at 10 μM). Inhibits the conversion of prednisone to prednisolone in human hepatocytes in vitro (EC50 = 15 nM) and has antidiabetic activity in vivo. Orally active.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 857290-04-1
Molekylær formel C18H14N4O2S
Tekniske data
Kemisk navn eller materiale PF 915275
Mængde 50 mg
Formel vægt 350.39
Renhed >98%
Molekylvægt (g/mol) 350.396