Få mere at vide
PF 5006739, Tocris Bioscience™
Potent CK1δ/ε inhibitor
Brand: Tocris Bioscience 6346/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PF 5006739 is a potent casein kinase 1δ/ε inhibitor (IC50 values are 3.9 and 17 nM, respectively). Mediates circadian rhythm phase-delaying effects in vivo. Attenuates opioid drug seeking behavior in addiction in vivo models.Tekniske data
| 4-[4-(4-Fluorophenyl)-1-[1-(3-isoxazolylmethyl)-4-piperidinyl]-1H-imidazol-5-yl]-2-pyrimidinamine | |
| 1293395-67-1 | |
| >98% | |
| 25 mg |