Få mere at vide
PF 5006739, Tocris Bioscience™
GREENER_CHOICE
Potent CK1δ/ε inhibitor
Brand:  Tocris Bioscience 6346/5
Encompass
Produktkode 15786409
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
PF 5006739 is a potent casein kinase 1δ/ε inhibitor (IC50 values are 3.9 and 17 nM, respectively). Mediates circadian rhythm phase-delaying effects in vivo. Attenuates opioid drug seeking behavior in addiction in vivo models.
Tekniske data
Tekniske data
4-[4-(4-Fluorophenyl)-1-[1-(3-isoxazolylmethyl)-4-piperidinyl]-1H-imidazol-5-yl]-2-pyrimidinamine
1293395-67-1
>98%
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner