Få mere at vide
Tocris Bioscience™ PF 429242
GREENER_CHOICE
Artikelnummer. 18084312
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18084312 10 mg
18026012 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18084312 Leverandør Tocris Bioscience™ Leverandørnr. 3354/10
Competitive inhibitor of SREBP site 1 protease
Reversible, competitive inhibitor of sterol regulatory element-binding protein (SREBP) site 1 protease (IC50 = 0.175 μM). Selective for site 1 protease against a panel of serine proteases. Inhibits rate of cholesterol synthesis in CHO cells (IC50 = 0.53 μM). Also displays antiviral activity. Cell permeable.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 947303-87-9
Molekylær formel C25H35N3O2
Tekniske data
Kemisk navn eller materiale PF 429242
Mængde 10 mg
Formel vægt 482.49
Renhed >97%
Molekylvægt (g/mol) 409.574