Få mere at vide
PF 06650833, Tocris Bioscience™
Potent and selective IRAK4 inhibitor
Brand: Tocris Bioscience 6373/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PF 06650833 is a potent and selective IRAK4 inhibitor (IC50 = 2 nM). Inhibits LPS-induced TNF-α secretion in vivo. Orally bioavailable.Tekniske data
| 1-[[(2S,3S,4S)-3-Ethyl-4-fluoro-5-oxo-2-pyrrolidinyl]methoxy]-7-methoxy-6-isoquinolinecarboxamide | |
| 1817626-54-2 | |
| >98% | |
| 5 mg |