Få mere at vide
Tocris Bioscience™ PF 06465469
GREENER_CHOICE
Artikelnummer. 30001555
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001555 10 mg
30001571 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001555 Leverandør Tocris Bioscience™ Leverandørnr. 4710/10
Potent ITK inhibitor; also inhibits BTK
Potent inhibitor of interleukin-2 inducible T cell kinase (ITK) (IC50 = 2 nM). Also exhibits inhibitory activity against Bruton's tyrosine kinase (BTK) (IC50 = 2 nM). Displays nanomolar potencies in a cell-based IP1 assay and human whole blood assay (IC50 values are 31 and 48 nM respectively).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1407966-77-1
Molekylær formel C30H33N7O2
Tekniske data
Kemisk navn eller materiale PF 06465469
Mængde 10 mg
Formel vægt 523.63
Renhed >95%
Molekylvægt (g/mol) 523.641