Få mere at vide
Tocris Bioscience™ PF 05175157
Acetyl-CoA carboxylase (ACC) 1 and 2 inhibitor
Brand: Tocris Bioscience™ 5790/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PF 05175157 is an acetyl-CoA carboxylase (ACC) 2 and 1 inhibitor (IC50 values are 45 and 98 nM, respectively). Reduces hepatic and skeletal muscle malonyl-CoA levels in rats. Also inhibits hepatic de novo lipogenesis and reduces whole body respiratory exchange ratio in rats. Orally bioavailable.Tekniske data
| 1,4-Dihydro-1′-[2-methyl-1H-benzimidazol-6-yl)carbonyl]-1-(1-methylethyl)-spiro[5H-indazole-5,4′-piperidin]-7(6H)-one | |
| 1301214-47-0 | |
| 10 mg |