Få mere at vide
PF 04628935, Tocris Bioscience™
Potent ghrelin receptor inverse agonist
Brand: Tocris Bioscience 6347/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.Tekniske data
| 1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone | |
| 1383719-97-8 | |
| >98% | |
| 25 mg |